2,4-Dichloropyridine 97%

Warning

L20040-03 L20040-03 L20040-06 L20040-06
AAAL20040-06EA 106.95 USD
AAAL20040-06 AAAL20040-03 CAAAAL20040-06 CAAAAL20040-03
2,4-Dichloropyridine 97%
2,4-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 189…190 °C
Melting Pt: –1 °C
Density: 1.37
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00955618
CAS Number: 26452-80-2
EINECS: 247-717-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR