6-Fluorosalicylaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-6-hydroxybenzaldehyde

L19900-MD L19900-03 L19900-MD L19900-03
AAAL19900-MDEA 82.79 USD
AAAL19900-MD CAAAAL19900-MD AAAL19900-03 CAAAAL19900-03
6-Fluorosalicylaldehyde 97%
6-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD01090996
CAS Number: 38226-10-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR