3'-Fluoro-4'-hydroxyacetophenone 98+%

Warning

Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

L19892-MD L19892-03 L19892-MD L19892-03
AAAL19892-03EA 160.95 USD
CAAAAL19892-03 AAAL19892-03 CAAAAL19892-MD AAAL19892-MD
3'-Fluoro-4'-hydroxyacetophenone 98+%
3'-Fluoro-4'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00215840
CAS Number: 403-14-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR