2,5-Difluorobenzenesulfonyl chloride 98%

Danger

Synonyms: 2,5-Difluorobenzene-1-sulfonyl chloride2,5-Difluorobenzene-1-sulphonyl chloride

L19890-06 L19890-06 L19890-03 L19890-03
AAAL19890-03EA 84.44 USD
CAAAAL19890-03 CAAAAL19890-06 AAAL19890-03 AAAL19890-06
2,5-Difluorobenzenesulfonyl chloride 98%
2,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 221 °C
Density: 1.580
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060682
CAS Number: 26120-86-5
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR