2,5-Bis(trifluoromethyl)benzenesulfonyl chloride 97%

Danger

Synonyms: 2,5-Bis(trifluoromethyl)benzene-1-sulfonyl chloride2,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride

L19878-03 L19878-03 L19878-06 L19878-06
AAAL19878-03EA 79.39 USD
CAAAAL19878-03 AAAL19878-03 CAAAAL19878-06 AAAL19878-06
2,5-Bis(trifluoromethyl)benzenesulfonyl chloride 97%
2,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₃ClF₆O₂S
MW: 312.62 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD03094042
CAS Number: 351003-22-0
EINECS: 000-000-0
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR