2-Chloro-3-picoline 98+%

Warning

Synonyms: 2-Chloro 3-methylpyridine2-Chloro-3-methylpyridine2-Chloro-β-picoline

L19861-14 L19861-06 L19861-06 L19861-14
AAAL19861-06EA 56.95 USD
CAAAAL19861-14 AAAL19861-06 CAAAAL19861-06 AAAL19861-14
2-Chloro-3-picoline 98+%
2-Chloro-3-picoline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 192…193 °C
Density: 1.170
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00234177
CAS Number: 18368-76-8
EINECS: 242-242-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR