2,3,5,6-Tetrafluorophenylboronic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(2,3,5,6-tetrafluorophenyl)borane
, (2,3,5,6-Tetrafluorophenyl)boronic acid
, 2,3,5,6-Tetrafluorobenzeneboronic acid
, (2,3,5,6-Tetrafluorophenyl)metaboric acid
L19826-03
L19826-03
AAAL19826-03EA
300.99
USD
CAAAAL19826-03
AAAL19826-03
2,3,5,6-Tetrafluorophenylboronic acid 99%
2,3,5,6-Tetrafluorophenylboronic acid
Formula:
C₆H₃BF₄O₂ MW: 193.89 g/mol Melting Pt: ca 140…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039310 CAS Number: 511295-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...