2-(Trifluoromethoxy)phenylboronic acid 98%

Warning

Synonyms: (2-(Trifluoromethoxy)phenyl)metaboric acidDihydroxy(2-(trifluoromethoxy)phenyl)borane2-Trifluoromethoxyphenylboronic acid2-(Trifluoromethoxy)benzeneboronic acid

L19774-03 L19774-MD L19774-MD L19774-03
AAAL19774-03EA 80.84 USD
AAAL19774-03 CAAAAL19774-03 AAAL19774-MD CAAAAL19774-MD
2-(Trifluoromethoxy)phenylboronic acid 98%
2-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD01320763
CAS Number: 175676-65-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR