1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

L19764-09 L19764-04 L19764-04 L19764-09
AAAL19764-04EA 68.99 USD
CAAAAL19764-04 AAAL19764-04 CAAAAL19764-09 AAAL19764-09
1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate 98+%
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
Density: 1.387
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00210008
CAS Number: 145022-44-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR