2-(Trifluoromethyl)benzenesulfonamide 97%

Warning

Synonyms: α,α,α-Trifluoro-o-toluenesulfonamideα,α,α-Trifluoro-o-toluenesulphonamide

L19717-06 L19717-03 L19717-06 L19717-03
AAAL19717-03EA 71.55 USD
AAAL19717-03 CAAAAL19717-06 CAAAAL19717-03 AAAL19717-06
2-(Trifluoromethyl)benzenesulfonamide 97%
2-(Trifluoromethyl)benzenesulphonamide
Formula: C₇H₆F₃NO₂S
MW: 225.19 g/mol
Melting Pt: 188…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00042420
CAS Number: 1869-24-5
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR