2-Fluoro-3-methoxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Fluoro-3-methoxyphenyl)boronic acid
, 2-Fluoro-3-methoxybenzeneboronic acid
L19655-03
L19655-06
L19655-06
L19655-03
AAAL19655-03EA
41.52
USD
CAAAAL19655-03
AAAL19655-03
CAAAAL19655-06
AAAL19655-06
2-Fluoro-3-methoxyphenylboronic acid 97%
2-Fluoro-3-methoxyphenylboronic acid
Formula:
C₇H₈BFO₃ MW: 169.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02094728 CAS Number: 352303-67-4 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...