5-Bromoquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

L19634-03 L19634-03 L19634-MD
AAAL19634-03EA 113.99 USD
AAAL19634-03 CAAAAL19634-MD CAAAAL19634-03
5-Bromoquinoline 97%
5-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 105…107 °C/1 mm
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00234481
CAS Number: 4964-71-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR