5-Bromoisoquinoline 98%

Warning

L19632-03 L19632-06 L19632-03 L19632-MD L19632-MD L19632-06
AAL19632-06EA 210.12 USD
CAAAAL19632-03 AAL19632-06 CAAAL19632-06 AAAL19632-03 AAAL19632-MD CAAAAL19632-MD
5-Bromoisoquinoline 98%
5-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 95…97 °C/0.1 mm
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD01646405
CAS Number: 34784-04-8
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR