3-Amino-2-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Nitropyridin-3-amine2-Nitropyridin-3-ylamine2-Nitro-3-pyridylamine

L19619-03 L19619-06 L19619-03 L19619-06
AAAL19619-03EA 164.65 USD
CAAAAL19619-03 CAAAAL19619-06 AAAL19619-03 AAAL19619-06
3-Amino-2-nitropyridine 97%
3-Amino-2-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00044102
CAS Number: 13269-19-7
EINECS: 236-260-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR