2,3-Dibromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19527-03 L19527-14 L19527-14 L19527-06 L19527-06 L19527-03
AAAL19527-03EA 43.56 USD
CAAAAL19527-06 AAAL19527-03 AAAL19527-14 AAAL19527-06 CAAAAL19527-14 CAAAAL19527-03
2,3-Dibromopyridine 97%
2,3-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 249…250 °C
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00234014
CAS Number: 13534-89-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR