2-Fluoronicotinic acid 97%

Warning

Synonyms: 2-Fluoro-3-pyridinecarboxylic acid2-Fluoropyridine-3-carboxylic acid

L19442-03 L19442-03 L19442-06 L19442-MD L19442-MD L19442-06
AAAL19442-03EA 75.77 USD
AAAL19442-03 CAAAAL19442-06 CAAAAL19442-MD AAAL19442-06 AAAL19442-MD CAAAAL19442-03
2-Fluoronicotinic acid 97%
2-Fluoronicotinic acid
Formula: C₆H₄FNO₂
Melting Pt: ca 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR