5-Bromotryptamine hydrochloride 98%

Supplier: Alfa Aesar
Warning

L19418-MD L19418-MD
AAAL19418-MDEA 80.52 USD
AAAL19418-MD CAAAAL19418-MD
5-Bromotryptamine hydrochloride 98%
5-Bromotryptamine hydrochloride
Formula: C₁₀H₁₂BrClN₂
MW: 275.58 g/mol
Melting Pt: ∼ 274 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00152022
CAS Number: 81868-12-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR