1-(2-Fluorophenyl)piperazine 99%

Danger

L19395-06 L19395-14 L19395-06
AAAL19395-06EA 39.38 USD
AAAL19395-06 AAAL19395-14 CAAAAL19395-06
1-(2-Fluorophenyl)piperazine 99%
1-(2-Fluorophenyl)piperazine
Formula: C₁₀H₁₃FN₂
MW: 180.23 g/mol
Boiling Pt: 150 °C/3 mm
Melting Pt: 44…48 °C
Density: 1.141
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040726
CAS Number: 1011-15-0
EINECS: 213-780-4
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR