1-(2,4-Difluorophenyl)piperazine 99%

Warning

L19393-06 L19393-03 L19393-03 L19393-14 L19393-06
AAAL19393-03EA 45.85 USD
AAAL19393-03 AAAL19393-14 CAAAAL19393-06 CAAAAL19393-03 AAAL19393-06
1-(2,4-Difluorophenyl)piperazine 99%
1-(2,4-Difluorophenyl)piperazine
Formula: C₁₀H₁₂F₂N₂
MW: 198.22 g/mol
Boiling Pt: 108…111 °C/0.2 mm
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00082588
CAS Number: 115761-79-0
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR