5-Fluoro-2-nitrobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

L19327-06 L19327-03 L19327-03 L19327-06
AAAL19327-03EA 82.59 USD
CAAAAL19327-03 AAAL19327-03 CAAAAL19327-06 AAAL19327-06
5-Fluoro-2-nitrobenzaldehyde 98%
5-Fluoro-2-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.11 g/mol
Boiling Pt: 153 °C/23 mm
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00153175
CAS Number: 395-81-3
EINECS: 206-903-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR