3-Bromo-2-chlorothiophene 97%

Warning

L19318-03 L19318-03 L19318-06 L19318-06
AAAL19318-03EA 46.34 USD
CAAAAL19318-03 AAAL19318-03 CAAAAL19318-06 AAAL19318-06
3-Bromo-2-chlorothiophene 97%
3-Bromo-2-chlorothiophene
Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 70 °C/9 mm
Density: 1.808
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043883
CAS Number: 40032-73-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR