(R)-(+)-1-(1-Naphthyl)ethylamine ≥99%, ee 98%, ChiPros®

Warning

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine(R)-1-(Naphthalen-1-yl)ethanamine

L19300-06 L19300-03 L19300-03 L19300-14 L19300-06
AAAL19300-03EA 59.58 USD
AAAL19300-03 AAAL19300-14 AAAL19300-06 CAAAAL19300-06 CAAAAL19300-03
(R)-(+)-1-(1-Naphthyl)ethylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 152…153 °C/11 mm
Density: 1.060
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR