2,4,5-Trifluoro-3-methoxybenzoic acid 98%

Warning

Synonyms: 3-Methoxy-2,4,5-trifluorobenzoic acid

L19237-03 L19237-06 L19237-06 L19237-03
AAAL19237-03EA 20.09 USD
CAAAAL19237-03 CAAAAL19237-06 AAAL19237-03 AAAL19237-06
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
2,4,5-Trifluoro-3-methoxybenzoic acid
Formula: C₈H₅F₃O₃
MW: 206,12 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00153201
CAS Number: 112811-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR