2,3,5-Trifluorobenzyl bromide 97%

Danger

Synonyms: α-Bromo-2,3,5-trifluorotoluene

L19191-03 L19191-03 L19191-06 L19191-06
AAAL19191-06EA 201.17 USD
AAAL19191-06 CAAAAL19191-06 AAAL19191-03 CAAAAL19191-03
2,3,5-Trifluorobenzyl bromide 97%
2,3,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Melting Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00083543
CAS Number: 226717-83-5
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR