3,4,5-Trifluorobenzoyl chloride 97%

Danger

L19188-MD L19188-06 L19188-MD L19188-03 L19188-06 L19188-03
AAAL19188-06EA 237.54 USD
AAAL19188-06 CAAAAL19188-MD AAAL19188-MD CAAAAL19188-03 AAAL19188-03 CAAAAL19188-06
3,4,5-Trifluorobenzoyl chloride 97%
3,4,5-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Melting Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00083526
CAS Number: 177787-26-7
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR