3-Fluoro-4-methylbenzyl alcohol 97%

Warning

Synonyms: (3-Fluoro-4-methylphenyl)methanol

L19170-03 L19170-03 L19170-MD L19170-MD
AAAL19170-03EA 97.38 USD
CAAAAL19170-MD AAAL19170-03 AAAL19170-MD CAAAAL19170-03
3-Fluoro-4-methylbenzyl alcohol 97%
3-Fluoro-4-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.16 g/mol
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD01631533
CAS Number: 192702-79-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR