3,4,5-Trifluorobenzoic acid 97%

Warning

L18939-06 L18939-03 L18939-06 L18939-03
AAAL18939-06EA 244.84 USD
AAAL18939-06 CAAAAL18939-06 AAAL18939-03 CAAAAL18939-03
3,4,5-Trifluorobenzoic acid 97%
3,4,5-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00074962
CAS Number: 121602-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR