Octyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Octyl isocyanate

L18746-06 L18746-06 L18746-03 L18746-03
AAAL18746-03EA 28.12 USD
CAAAAL18746-06 AAAL18746-03 AAAL18746-06 CAAAAL18746-03
Octyl isocyanate 97%
Octyl isocyanate
Formula: C₉H₁₇NO
MW: 155.24 g/mol
Boiling Pt: 200…204 °C
Density: 0.880
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013887
CAS Number: 3158-26-7
EINECS: 221-598-7
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR