4-Pyridineboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

L17854-03 L17854-MD L17854-03 L17854-MD
AAAL17854-MDEA 43.99 USD
CAAAAL17854-MD AAAL17854-MD CAAAAL17854-03 AAAL17854-03
4-Pyridineboronic acid pinacol ester 98%
4-Pyridineboronic acid pinacol ester
Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD01319051
CAS Number: 181219-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR