3-Iodothiophene 96% stabilized

Warning


Stabilization: Stabilised with copper

L17581-06 L17581-03 L17581-06 L17581-03
AAAL17581-03EA 76.25 USD
CAAAAL17581-03 AAAL17581-03 CAAAAL17581-06 AAAL17581-06
3-Iodothiophene 96% stabilized
3-Iodothiophene
Formula: C₄H₃IS
MW: 210.04 g/mol
Boiling Pt: 77 °C/11 mm
Density: 2.060
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037901
CAS Number: 10486-61-0
EINECS: 234-009-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR