4-Benzyloxyindole 99%

Warning

L17147-MD L17147-03 L17147-03 L17147-MD
AAAL17147-MDEA 41.35 USD
AAAL17147-MD CAAAAL17147-MD AAAL17147-03 CAAAAL17147-03
4-Benzyloxyindole 99%
4-Benzyloxyindole
Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00047200
CAS Number: 20289-26-3
EINECS: 243-690-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR