(S)-(-)-1-(2-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®

Warning

Synonyms: (S)-1-(Naphthalen-2-yl)ethanamine

L17024-03 L17024-03 L17024-06 L17024-06
AAAL17024-06EA 202.15 USD
CAAAAL17024-06 AAAL17024-06 CAAAAL17024-03 AAAL17024-03
(S)-(-)-1-(2-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-1-(2-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 90 °C/0.15 mm
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00085366
CAS Number: 3082-62-0
EINECS: 608-543-0
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR