2,2,2-Trifluoroethyl acetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid (2,2,2-trifluoroethyl) ester

L16871-22 L16871-22 L16871-14 L16871-14
AAAL16871-22EA 226.99 USD
AAAL16871-22 CAAAAL16871-22 AAAL16871-14 CAAAAL16871-14
2,2,2-Trifluoroethyl acetate 97%
2,2,2-Trifluoroethyl acetate
Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 78 °C
Density: 1.258
Flash Pt: 11 °C(51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00198018
CAS Number: 406-95-1
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR