1-(2-Chloroethyl)-4-fluorobenzene 97%

Warning

L16719-06 L16719-03 L16719-06 L16719-03
AAAL16719-06EA 329.54 USD
AAAL16719-06 AAAL16719-03 CAAAAL16719-06 CAAAAL16719-03
1-(2-Chloroethyl)-4-fluorobenzene 97%
1-(2-Chloroethyl)-4-fluorobenzene
Formula: C₈H₈ClF
MW: 158.6 g/mol
Boiling Pt: 98…99 °C/26 mm
Density: 1.507
Storage Temperature: Ambient
MDL Number: MFCD00155000
CAS Number: 332-43-4
EINECS: 206-364-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR