2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,3,3,5,5,7,7-Octamethylcyclotetrasilazane

L16563-06 L16563-14 L16563-06 L16563-14
AAAL16563-14EA 71.39 USD
CAAAAL16563-06 AAAL16563-14 CAAAAL16563-14 AAAL16563-06
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane 97%
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane
Formula: C₈H₂₈N₄Si₄
MW: 292.68 g/mol
Boiling Pt: 224…225 °C
Melting Pt: 94…98 °C
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046123
CAS Number: 1020-84-4
EINECS: 213-817-4
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR