Diethylsilane ≥97%

Supplier: Thermo Scientific Chemicals
Danger

L16468-14 L16468-14 L16468-06 L16468-06
AAAL16468-06EA 75.31 USD
AAAL16468-06 CAAAAL16468-14 AAAL16468-14 CAAAAL16468-06
Diethylsilane ≥97%
Diethylsilane
Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 56 °C
Melting Pt: –132 °C
Density: 0.684
Flash Pt: -20 °C(-4 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039877
CAS Number: 542-91-6
EINECS: 208-834-9
UN: 1993
ADR: "3",III
Merck Index: 13,03154

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR