2-Bromo-5-nitropyridine 98+%

Warning

L15661-03 L15661-03 L15661-06 L15661-06
AAAL15661-03EA 30.28 USD
CAAAAL15661-03 AAAL15661-03 CAAAAL15661-06 AAAL15661-06
2-Bromo-5-nitropyridine 98+%
2-Bromo-5-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Boiling Pt: 145…147 °C/10 mm
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR