Zirconium tetrachloride 98% cont. 1-2% hafnium(IV) chloride

Danger

Synonyms: Zirconium (IV) chloride

L14891-18 L14891-30 L14891-18 L14891-30
AAAL14891-30EA 81.87 USD
AAAL14891-30 AAAL14891-18 CAAAAL14891-18 CAAAAL14891-30
Zirconium tetrachloride 98% cont. 1-2% hafnium(IV) chloride
Zirconium tetrachloride
Formula: Cl₄Zr
MW: 233.03 g/mol
Melting Pt: 331 °C (subl.)
Density: 2.803
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: "8",III
Merck Index: 14,10174

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR