Lactic acid 80-85% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydroxypropionic acid
, 2-Hydroxypropanoic acid
, DL-Lactic acid
L14259-0E
L14259-0E
L14259-0B
L14259-30
AAAL14259-0EEA
236.27
USD
CAAAAL14259-0E
AAAL14259-0E
AAL14259-0B
AAL14259-30
Lactic acid 80-85% in aqueous solution
Lactic acid
Formula:
H₃CCH(OH)COOH MW: 90.08 g/mol Boiling Pt: 122 °C/15 mm Density: 1.209 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004520 CAS Number: 50-21-5 EINECS: 200-018-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...