2'-Hydroxychalcone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Hydroxyphenyl)-3-phenylprop-2-en-1-one2'-Hydroxybenzylidenacetophenone1-(2-Hydroxyphenyl)-3-phenyl-2-propenone

L13937-06 L13937-14
AAAL13937-06EA 33.19 USD
AAAL13937-06 AAAL13937-14
2'-Hydroxychalcone 98+%
2'-Hydroxychalcone
Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00016441
CAS Number: 1214-47-7
EINECS: 214-928-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR