α-Bromo-2,5-dichlorotoluene 97%

Danger

Synonyms: 2-(Bromomethyl)-1,4-dichlorobenzene2,5-Dichlorobenzyl bromide

L13775-06 L13775-03 L13775-03 L13775-14 L13775-14 L13775-06
AAAL13775-03EA 28.48 USD
CAAAAL13775-03 CAAAAL13775-14 AAAL13775-03 AAAL13775-14 CAAAAL13775-06 AAAL13775-06
α-Bromo-2,5-dichlorotoluene 97%
α-Bromo-2,5-dichlorotoluene
Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00236034
CAS Number: 85482-13-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR