2-Nitrobenzyl chloride 98+%

Danger

Synonyms: 2-Nitrobenzyl chloride1-(Chloromethyl)-2-nitrobenzeneα-Chloro-o-nitrotoluenealpha-Chloro-2-nitrotoluene

L13599-14 L13599-14
AAAL13599-14EA 227.55 USD
AAAL13599-14 CAAAAL13599-14
2-Nitrobenzyl chloride 98+%
2-Nitrobenzyl chloride
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 127…133 °C/10 mm
Melting Pt: 47…51 °C
Density: 1.556
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007185
CAS Number: 612-23-7
EINECS: 210-300-5
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR