3-Chloroanilinium chloride 97%

Danger

Synonyms: 3-Chloroaniline hydrochloride

L13484-14 L13484-22 L13484-14 L13484-22
AAAL13484-22EA 73.84 USD
AAAL13484-22 CAAAAL13484-22 AAAL13484-14 CAAAAL13484-14
3-Chloroanilinium chloride 97%
3-Chloroanilinium chloride
Formula: C₆H₇Cl₂N
Melting Pt: ca 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035485
CAS Number: 141-85-5
EINECS: 205-506-7
UN: 2018
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR