4-Butylbenzaldehyde 97%

L13438-03 L13438-03 L13438-06
AAAL13438-03EA 44.11 USD
CAAAAL13438-06 AAAL13438-03 CAAAAL13438-03
4-Butylbenzaldehyde 97%
4-Butylbenzaldehyde
Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 96…98 °C/5 mm
Melting Pt: –43 °C
Density: 0.970
Storage Temperature: Ambient
MDL Number: MFCD00040751
CAS Number: 1200-14-2
EINECS: 214-851-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR