N,N-Dibutylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

L13390-06 L13390-06
AAAL13390-06EA 26.39 USD
AAAL13390-06 CAAAAL13390-06
N,N-Dibutylaniline 97%
N,N-Dibutylaniline
Formula: (C₆H₅)N(CH₂CH₂CH₂CH₃)₂
MW: 205.34 g/mol
Boiling Pt: 269…270 °C
Melting Pt: –32 °C
Density: 0.906
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035799
CAS Number: 613-29-6
EINECS: 210-335-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR