1,1,3-Triethoxypropane 98+%

Warning

L13011-06 L13011-06 L13011-14 L13011-14
AAAL13011-14EA 104.51 USD
AAAL13011-14 CAAAAL13011-06 CAAAAL13011-14 AAAL13011-06
1,1,3-Triethoxypropane 98+%
1,1,3-Triethoxypropane
Formula: C₉H₂₀O₃
Boiling Pt: 184…186 °C
Density: 0.898
Flash Pt: 53 °C(127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009241
CAS Number: 7789-92-6
EINECS: 232-193-4
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR