3,6-Dichloroanthranilic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3,6-dichlorobenzoic acid

L12850-06 L12850-06 L12850-03 L12850-03
AAAL12850-06EA 197.99 USD
CAAAAL12850-03 AAAL12850-06 AAAL12850-03 CAAAAL12850-06
3,6-Dichloroanthranilic acid 95%
3,6-Dichloroanthranilic acid
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00052390
CAS Number: 3032-32-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR