2,4-Dimethoxyphenyl isothiocyanate 97%

Warning

Synonyms: 1,3-Dimethoxy-4-isothiocyanatobenzene2,4-Dimethoxyphenylisothiocyanate

L12584-03 L12584-06 L12584-06 L12584-03
AAAL12584-03EA 26.25 USD
AAAL12584-03 CAAAAL12584-03 AAAL12584-06 CAAAAL12584-06
2,4-Dimethoxyphenyl isothiocyanate 97%
2,4-Dimethoxyphenyl isothiocyanate
Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Boiling Pt: 166…167 °C/6 mm
Melting Pt: 51…53 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041058
CAS Number: 33904-03-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR