2-Hydroxythiophenol 97%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Hydroxythiophenol2-Mercaptophenol1-Hydroxy-2-mercaptobenzene2-Hydroxyphenyl mercaptan2-Hydroxybenzene-1-thiol2-Hydroxybenzenethiol

L12519-06 L12519-06
AAAL12519-06EA 172.99 USD
AAAL12519-06 CAAAAL12519-06
2-Hydroxythiophenol 97%, Technical Grade
2-Hydroxythiophenol
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 217 °C
Melting Pt: 19 °C
Density: 1.220
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040447
CAS Number: 1121-24-0
EINECS: 214-326-8
UN: 2927
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR