2,4,6-Trimethylphenyl isothiocyanate 96%

Warning

Synonyms: 1-Isothiocyanato-2,4,6-trimethylbenzene2,4,6-Trimethylphenylisothiocyanate

L12294-03 L12294-03 L12294-06 L12294-06
AAAL12294-06EA 102.02 USD
AAAL12294-06 CAAAAL12294-06 AAAL12294-03 CAAAAL12294-03
2,4,6-Trimethylphenyl isothiocyanate 96%
2,4,6-Trimethylphenyl isothiocyanate
Formula: C₁₀H₁₁NS
MW: 177.27 g/mol
Boiling Pt: 144…145 °C/10 mm
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00022055
CAS Number: 6095-82-5
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR